1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrothiophen-2-yl)pyrimidine-2,4(1H,3H)-dione ≥97%

Supplier: Ambeed
Warning

Synonyms: 4′-Thiouridine

A420433-10MG A420433-5MG A420433-50MG
77616-822EA 428.48 USD
77616-822 77616-824 77746-845
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrothiophen-2-yl)pyrimidine-2,4(1H,3H)-dione ≥97%
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrothiophen-2-yl)pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₅S
MW: 260.27 g/mol
MDL Number: MFCD22666335
CAS Number: 6741-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR