(5-Chlorobenzofuran-2-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A429282-1G A429282-100MG A429282-250MG A429282-5G
77572-332EA 168 USD
77572-332 77892-891 77892-892 77892-893
(5-Chlorobenzofuran-2-yl)boronic acid ≥98%
(5-Chlorobenzofuran-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₆BClO₃
MDL Number: MFCD18383028
CAS Number: 223576-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR