DBCO-PEG4-NHS Ester ≥95%

Supplier: Ambeed
Warning

Synonyms: Dibenzocyclooctyne-PEG4-succinimidyl Ester, 2,5-Dioxo-1-pyrrolidinyl 20-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)-17,20-dioxo-4,7,10,13-tetraoxa-16-azaeicosanoate, DBCO-PEG4-succinimidyl Ester, ADIBO-PEG4-NHS, DIBACA-PEG4-NHS

A420011-250MG A420011-1G A420011-50MG
77746-843EA 492 USD
77746-843 77746-842 77746-844
DBCO-PEG4-NHS Ester ≥95%
DBCO-PEG4-NHS Ester

Precaución: for Research Use Only

Formula: C₃₄H₃₉N₃O₁₀
MDL Number: MFCD26793797
CAS Number: 1427004-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR