(3,4-Diethoxyphenyl)acetonitrile ≥95%

Supplier: Ambeed
Warning

A419858-1G A419858-25G A419858-5G
77879-918EA 40 USD
77879-918 77879-919 77879-920
(3,4-Diethoxyphenyl)acetonitrile ≥95%
(3,4-Diethoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₂
MDL Number: MFCD00060715
CAS Number: 27472-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR