Tricyclohexylphosphine[3-phenyl-1H-inden-1-ylidene][1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]ruthenium(II) dichloride ≥98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
[1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro-(3-phenyl-1H-inden-1-ylidene)(tricyclohexylphosphine)ruthenium(II)
A419773-250MG
A419773-1G
77684-175EA
95
USD
77684-175
77886-109
Tricyclohexylphosphine[3-phenyl-1H-inden-1-ylidene][1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]ruthenium(II) dichloride ≥98%, Ambeed.Inc
Tricyclohexylphosphine[3-phenyl-1H-inden-1-ylidene][1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]ruthenium(II) dichloride
Precaución: For research use only.
Formula:
C₅₄H₆₉Cl₂N₂PRu |
MDL Number:
MFCD08667892 CAS Number: 536724-67-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...