Tetraphenyl (propane-2,2-diylbis(4,1-phenylene)) bis(phosphate) ≥97%

Supplier: Ambeed
Warning

A429031-100G A429031-25G
77878-182EA 23 USD
77878-182 77878-183
Tetraphenyl (propane-2,2-diylbis(4,1-phenylene)) bis(phosphate) ≥97%
Tetraphenyl (propane-2,2-diylbis(4,1-phenylene)) bis(phosphate)

Precaución: for Research Use Only

Formula: C₃₉H₃₄O₈P₂
CAS Number: 5945-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR