(R)-3-Fluoropyrrolidine ≥98%

Supplier: Ambeed
Danger

A428904-5G A428904-250MG
77594-332EA 410 USD
77594-332 77890-877
(R)-3-Fluoropyrrolidine ≥98%
(R)-3-Fluoropyrrolidine

Precaución: for Research Use Only

Formula: C₄H₈FN
MDL Number: MFCD06738462
CAS Number: 679431-51-7
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR