Kavain (+/-) ≥99%

Supplier: Ambeed
Warning

Synonyms: (±)-4-Methoxy-6-styryl-5,6-dihydro-α-pyrone, (+)-Kavain, (R,E)-4-Methoxy-6-styryl-5,6-dihydro-2H-pyran-2-one

A428642-1MG A428642-10MG A428642-5MG
77601-848EA 33.83 USD
77601-848 77668-912 77668-914
Kavain (+/-) ≥99%
Kavain (+/-)

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₃
MW: 230.3 g/mol
MDL Number: MFCD00467093
CAS Number: 500-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR