6-(Aminomethyl)morpholin-3-one ≥95%
Supplier: Ambeed
|
Danger
|
A428306-100MG
A428306-1G
A428306-250MG
77884-107EA
78
USD
77884-107
77884-108
77884-109
6-(Aminomethyl)morpholin-3-one ≥95%
6-(Aminomethyl)morpholin-3-one
Precaución: for Research Use Only
Formula:
C₅H₁₀N₂O₂ |
MDL Number:
MFCD18071974 CAS Number: 793644-35-6 UN: 3259 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...