UM-164 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-[[6-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-methyl-4-pyrimidinyl]amino]-N-[2-methyl-5-[[3-(trifluoromethyl)benzoyl]amino]phenyl]-5-thiazolecarboxamide, 2-((6-(4-(2-Hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)-N-(2-methyl-5-(3-(trifluoromethyl)benzamido)phenyl)thiazole-5-carboxamide

A427983-10MG A427983-1MG A427983-5MG
77594-326EA 115.83 USD
77594-326 77594-328 77594-330
UM-164 ≥98%
UM-164

Precaución: for Research Use Only

Formula: C₃₀H₃₁F₃N₈O₃S
MW: 640.68 g/mol
MDL Number: MFCD31692378
CAS Number: 903564-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR