(S)-1-(2-Chlorophenyl)ethanamine ≥95%

Supplier: Ambeed
Warning

A427828-100MG A427828-250MG
77886-135EA 75 USD
77886-135 77886-136
(S)-1-(2-Chlorophenyl)ethanamine ≥95%
(S)-1-(2-Chlorophenyl)ethanamine

Precaución: for Research Use Only

Formula: C₈H₁₀ClN
MDL Number: MFCD06761892
CAS Number: 68285-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR