[RuCl(p-cymene)((R)-binap)]Cl ≥98%

Supplier: Ambeed
Danger

A427739-5G A427739-1G
77686-172EA 174 USD
77686-172 77686-171
[RuCl(p-cymene)((R)-binap)]Cl ≥98%
[RuCl(p-cymene)((R)-binap)]Cl

Precaución: for Research Use Only

Formula: C₅₄H₄₆Cl₂P₂Ru
MDL Number: MFCD00134456
CAS Number: 145926-28-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR