5-Methoxy-2-pyridineboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (5-Methoxypyridin-2-yl)boronic acid

A427627-100MG A427627-1G
77572-328EA 110 USD
77572-328 77872-686
5-Methoxy-2-pyridineboronic acid ≥98%
5-Methoxy-2-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₃
MDL Number: MFCD08752653
CAS Number: 1142944-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR