(R)-N-(1-(3-(4-Ethoxyphenyl)-4-oxo-3,4-dihydropyrido[2,3-d]pyrimidin-2-yl)ethyl)-N-(pyridin-3-ylmethyl)-2-(4-(trifluoromethoxy)phenyl)acetamide 98
Supplier: Ambeed
|
Warning
|
Synonyms:
AMG 487
A427101-5MG
A427101-1MG
A427101-100MG
A427101-10MG
77746-863EA
118
USD
77746-863
77746-862
77746-860
77746-861
(R)-N-(1-(3-(4-Ethoxyphenyl)-4-oxo-3,4-dihydropyrido[2,3-d]pyrimidin-2-yl)ethyl)-N-(pyridin-3-ylmethyl)-2-(4-(trifluoromethoxy)phenyl)acetamide 98
(R)-N-(1-(3-(4-Ethoxyphenyl)-4-oxo-3,4-dihydropyrido[2,3-d]pyrimidin-2-yl)ethyl)-N-(pyridin-3-ylmethyl)-2-(4-(trifluoromethoxy)phenyl)acetamide
Precaución: for Research Use Only
Formula:
C₃₂H₂₈F₃N₅O₄ MW: 603.59 g/mol |
MDL Number:
MFCD11111772 CAS Number: 473719-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...