(S)-Quinuclidin-3-yl (2-(2-(4-fluorophenyl)thiazol-4-yl)propan-2-yl)carbamate ≥98%

Supplier: Ambeed
Warning

Synonyms: Ibiglustat

A417980-100MG A417980-10MG A417980-2MG A417980-5MG
77686-152EA 491 USD
77686-152 77686-153 77686-154 77686-155
(S)-Quinuclidin-3-yl (2-(2-(4-fluorophenyl)thiazol-4-yl)propan-2-yl)carbamate ≥98%
(S)-Quinuclidin-3-yl (2-(2-(4-fluorophenyl)thiazol-4-yl)propan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₂₀H₂₄FN₃O₂S
MW: 389.49 g/mol
MDL Number: MFCD28502073
CAS Number: 1401090-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR