2-(Pyridin-3-yl)naphtho[2,3-d]oxazole-4,9-dione ≥99%

Supplier: Ambeed
Warning

Synonyms: SJB3-019A

A426930-250MG A426930-50MG A426930-100MG
77748-862EA 703 USD
77748-862 77748-863 77748-861
2-(Pyridin-3-yl)naphtho[2,3-d]oxazole-4,9-dione ≥99%
2-(Pyridin-3-yl)naphtho[2,3-d]oxazole-4,9-dione

Precaución: for Research Use Only

Formula: C₁₆H₈N₂O₃
MW: 276.25 g/mol
MDL Number: MFCD26960956
CAS Number: 2070015-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR