(2S)-Benzyl 2-(((((2R,3R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-4,4-difluoro-3-hydroxytetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)amino)propanoate 98+%

Supplier: Ambeed
Warning

Synonyms: Acelarin

A426804-50MG A426804-10MG
77748-860EA 452 USD
77748-860 77748-859
(2S)-Benzyl 2-(((((2R,3R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-4,4-difluoro-3-hydroxytetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)amino)propanoate 98+%
(2S)-Benzyl 2-(((((2R,3R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-4,4-difluoro-3-hydroxytetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)amino)propanoate

Precaución: for Research Use Only

Formula: C₂₅H₂₇F₂N₄O₈P
MW: 580.47 g/mol
MDL Number: MFCD27987942
CAS Number: 840506-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR