Boc-(S)-3-amino-4-(2-naphthyl)butyric acid ≥98%

Supplier: Ambeed
Warning

A417194-1G A417194-100MG A417194-250MG
77626-834EA 246.17 USD
77626-834 77878-169 77878-170
Boc-(S)-3-amino-4-(2-naphthyl)butyric acid ≥98%
Boc-(S)-3-amino-4-(2-naphthyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₉H₂₃NO₄
MDL Number: MFCD01076284
CAS Number: 219297-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR