4-Cyclopropyl-1,3-thiazole-2-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A426399-100MG A426399-250MG
77882-114EA 50 USD
77882-114 77882-115
4-Cyclopropyl-1,3-thiazole-2-carboxylic acid ≥95%
4-Cyclopropyl-1,3-thiazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₂S
MDL Number: MFCD07377487
CAS Number: 1083274-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR