3-Bromo-2-chloro-5-fluorobenzoic acid ≥97%

Supplier: Ambeed
Warning

A426122-5G A426122-1G
77594-320EA 75.85 USD
77594-320 77616-844
3-Bromo-2-chloro-5-fluorobenzoic acid ≥97%
3-Bromo-2-chloro-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₂
MDL Number: MFCD28166248
CAS Number: 1805210-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR