Methyl 7-bromo-1H-indazole-5-carboxylate ≥95%

Supplier: Ambeed
Warning

A425763-1G A425763-100MG
77613-316EA 64.89 USD
77613-316 77892-880
Methyl 7-bromo-1H-indazole-5-carboxylate ≥95%
Methyl 7-bromo-1H-indazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MDL Number: MFCD22421936
CAS Number: 1427460-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR