Octakis(6-Bromo-6-Deoxy)-γ-Cyclodextrin ≥98%

Supplier: Ambeed
Warning

Synonyms: Octakis-(6-bromo-6-deoxy)-γ-cyclodextrin

A416532-250MG A416532-100MG A416532-1G
77613-276EA 148.32 USD
77613-276 77686-149 77686-150
Octakis(6-Bromo-6-Deoxy)-γ-Cyclodextrin ≥98%
Octakis(6-Bromo-6-Deoxy)-γ-Cyclodextrin

Precaución: for Research Use Only

Formula: C₄₈H₇₂Br₈O₃₂
MW: 1800.29 g/mol
CAS Number: 53784-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR