Fmoc-3-iodo-L-Tyrosine ≥97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-3-iodo-Tyr-OH, Fmoc-Tyr(3-I)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-hydroxy-3-iodophenyl)propanoic acid

A425169-25G A425169-1G A425169-5G
77572-320EA 661 USD
77572-320 77900-791 77901-560
Fmoc-3-iodo-L-Tyrosine ≥97%
Fmoc-3-iodo-L-Tyrosine

Precaución: for Research Use Only

Formula: C₂₄H₂₀INO₅
MDL Number: MFCD00235906
CAS Number: 134486-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR