2-Amino-4,6-difluorobenzonitrile ≥98%
Supplier: Ambeed
|
Warning
|
A424613-1G
A424613-5G
77594-312EA
68.68
USD
77594-312
77684-183
2-Amino-4,6-difluorobenzonitrile ≥98%
2-Amino-4,6-difluorobenzonitrile
Precaución: For research use only.
Formula:
C₇H₄F₂N₂ MW: 154.117 g/mol |
MDL Number:
MFCD13178245 CAS Number: 190011-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...