Taurodeoxycholic acid sodium salt hydrate ≥95%

Supplier: Ambeed
Warning

A415821-10G A415821-1G A415821-25G A415821-5G
77613-274EA 106.09 USD
77613-274 77673-016 77673-018 77673-020
Taurodeoxycholic acid sodium salt hydrate ≥95%
Taurodeoxycholic acid sodium salt hydrate

Precaución: for Research Use Only

Formula: C₂₆H₄₄NNaO₆S.xH₂O
MDL Number: MFCD00149238
CAS Number: 207737-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR