(2R,3R,4S,5S,6R)-2-(Cinnamyloxy)-6-((((2S,3R,4S,5S)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol 99+%

Supplier: Ambeed
Warning

Synonyms: Rosavin

A424145-1MG A424145-5MG
77668-896EA 44 USD
77668-896 77668-898
(2R,3R,4S,5S,6R)-2-(Cinnamyloxy)-6-((((2S,3R,4S,5S)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol 99+%
(2R,3R,4S,5S,6R)-2-(Cinnamyloxy)-6-((((2S,3R,4S,5S)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol

Precaución: for Research Use Only

Formula: C₂₀H₂₈O₁₀
MDL Number: MFCD03427685
CAS Number: 84954-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR