Methyl 3-(bromomethyl)-2-fluorobenzoate ≥95%

Supplier: Ambeed
Danger

A415353-1G A415353-250MG
77594-278EA 295.2 USD
77594-278 77594-280
Methyl 3-(bromomethyl)-2-fluorobenzoate ≥95%
Methyl 3-(bromomethyl)-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.0610 g/mol
MDL Number: MFCD25959973
CAS Number: 171670-20-5
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR