5-(4-Bromophenyl)pentanoic acid ≥95%

Supplier: Ambeed
Warning

A414751-250MG A414751-100MG
77598-476EA 56.38 USD
77598-476 77783-404
5-(4-Bromophenyl)pentanoic acid ≥95%
5-(4-Bromophenyl)pentanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MDL Number: MFCD09890955
CAS Number: 22647-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR