6-(Trifluoromethyl)pyridine-3-sulfonyl chloride ≥98%

Supplier: Ambeed
Danger

A415255-5G A415255-10G
77882-090EA 357 USD
77882-090 77882-089
6-(Trifluoromethyl)pyridine-3-sulfonyl chloride ≥98%
6-(Trifluoromethyl)pyridine-3-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃ClF₃NO₂S
MDL Number: MFCD10699112
CAS Number: 959996-58-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR