4-[(N,N-Dimethylamino)methyl]benzeneboronic acid pinacol ester hydrochloride ≥98%

Supplier: Ambeed
Warning

A414463-250MG A414463-5G A414463-1G
77890-854EA 28 USD
77890-854 77890-855 77890-853
4-[(N,N-Dimethylamino)methyl]benzeneboronic acid pinacol ester hydrochloride ≥98%
4-[(N,N-Dimethylamino)methyl]benzeneboronic acid pinacol ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₄BNO₂·ClH
MDL Number: MFCD07771993
CAS Number: 1073371-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR