Ethyl 2,4-dichloropyrrolo[2,1-f]-[1,2,4]triazine-6-carboxylate ≥95%

Supplier: Ambeed
Warning

A414402-1G A414402-250MG
77594-276EA 994.25 USD
77594-276 77900-671
Ethyl 2,4-dichloropyrrolo[2,1-f]-[1,2,4]triazine-6-carboxylate ≥95%
Ethyl 2,4-dichloropyrrolo[2,1-f]-[1,2,4]triazine-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇Cl₂N₃O₂
MDL Number: MFCD21642058
CAS Number: 1363381-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR