(3aR,5R,6S,7R,7aR)-5-(Hydroxymethyl)-2-methyl-5,6,7,7a-tetrahydro-3aH-pyrano[3,2-d]thiazole-6,7-diol ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: NAG-thiazoline

A415003-250MG A415003-100MG A415003-1G A415003-50MG
77613-270EA 767.35 USD
77613-270 77684-166 77684-167 77684-168
(3aR,5R,6S,7R,7aR)-5-(Hydroxymethyl)-2-methyl-5,6,7,7a-tetrahydro-3aH-pyrano[3,2-d]thiazole-6,7-diol ≥95%, Ambeed.Inc
(3aR,5R,6S,7R,7aR)-5-(Hydroxymethyl)-2-methyl-5,6,7,7a-tetrahydro-3aH-pyrano[3,2-d]thiazole-6,7-diol

Precaución: For research use only.

Formula: C₈H₁₃NO₄S
MW: 219.26 g/mol
MDL Number: MFCD22376563
CAS Number: 179030-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR