4-(2-(((6R,7R)-2-Carboxy-7-((Z)-2-(ethoxyimino)-2-(5-(phosphonoamino)-1,2,4-thiadiazol-3-yl)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)thio)thiazol-4-yl)-1-methylpyridin-1-ium acetate ≥98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Ceftaroline fosamil
A414202-1MG
A414202-5MG
A414202-10MG
A414202-50MG
77613-266EA
5.15
USD
77613-266
77613-268
77684-164
77684-165
4-(2-(((6R,7R)-2-Carboxy-7-((Z)-2-(ethoxyimino)-2-(5-(phosphonoamino)-1,2,4-thiadiazol-3-yl)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)thio)thiazol-4-yl)-1-methylpyridin-1-ium acetate ≥98%
4-(2-(((6R,7R)-2-Carboxy-7-((Z)-2-(ethoxyimino)-2-(5-(phosphonoamino)-1,2,4-thiadiazol-3-yl)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)thio)thiazol-4-yl)-1-methylpyridin-1-ium acetate
Precaución: For research use only.
Formula:
C₂₄H₂₅N₈O₁₀PS₄ MW: 744.74 g/mol |
MDL Number:
MFCD19443689 CAS Number: 400827-46-5 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...