(1H-1,2,3-Triazol-5-yl)methanamine hydrochloride ≥96%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A406363-1G
A406363-250MG
77684-145EA
199
USD
77684-145
77684-146
(1H-1,2,3-Triazol-5-yl)methanamine hydrochloride ≥96%, Ambeed.Inc
(1H-1,2,3-Triazol-5-yl)methanamine hydrochloride
Precaución: For research use only.
Formula:
C₃H₇ClN₄ MW: 134.57 g/mol |
MDL Number:
MFCD06738997 CAS Number: 1009101-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...