2-Amino-2-methylbutanoic acid ≥98%

Supplier: Ambeed
Warning

A413670-100MG A413670-1G
77886-101EA 29 USD
77886-101 77886-102
2-Amino-2-methylbutanoic acid ≥98%
2-Amino-2-methylbutanoic acid

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MDL Number: MFCD00272968
CAS Number: 595-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR