SNS-314 mesylate ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Chlorophenyl)-3-(5-(2-(thieno[3,2-d]pyrimidin-4-ylamino)ethyl)thiazol-2-yl)urea methanesulphonate, N-(3-Chlorophenyl)-N′-[5-[2-(thieno[3,2-d]pyrimidin-4-ylamino)ethyl]-2-thiazolyl]urea methanesulphonate, (3-chlorophenyl)-3-(5-(2-(thieno[3,2-d]pyrimidin-4-ylamino)ethyl)thiazol-2-yl)urea mesyalte

A405855-5MG A405855-10MG
77748-821EA 66 USD
77748-821 77748-820
SNS-314 mesylate ≥98%
SNS-314 mesylate

Precaución: for Research Use Only

Formula: C₁₈H₁₅ClN₆OS₂·CH₄O₃S
MW: 527.04 g/mol
MDL Number: MFCD16621142
CAS Number: 1146618-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR