6-Bromo-3-pyridazinecarboxylic acid ≥95%

Supplier: Ambeed
Warning

A405531-1G A405531-250MG
77572-256EA 389.34 USD
77572-256 77626-818
6-Bromo-3-pyridazinecarboxylic acid ≥95%
6-Bromo-3-pyridazinecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₂
MW: 202.99 g/mol
MDL Number: MFCD06200883
CAS Number: 65202-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR