(S)-7-Hydroxy-2-(4-hydroxy-3,5-bis(3-methylbut-2-en-1-yl)phenyl)-8-(3-methylbut-2-en-1-yl)chroman-4-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Sophoranone

A405496-1MG A405496-5MG
77616-762EA 91.67 USD
77616-762 77748-818
(S)-7-Hydroxy-2-(4-hydroxy-3,5-bis(3-methylbut-2-en-1-yl)phenyl)-8-(3-methylbut-2-en-1-yl)chroman-4-one ≥98%
(S)-7-Hydroxy-2-(4-hydroxy-3,5-bis(3-methylbut-2-en-1-yl)phenyl)-8-(3-methylbut-2-en-1-yl)chroman-4-one

Precaución: for Research Use Only

Formula: C₃₀H₃₆O₄
MDL Number: MFCD29048556
CAS Number: 23057-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR