N-(9-Fluorenylmethoxycarbonyl)-5-aminopentan-1-ol ≥95%

Supplier: Ambeed
Warning

Synonyms: (9H-Fluoren-9-yl)methyl (5-hydroxypentyl)carbamate

A412875-1G A412875-250MG A412875-5G
77616-798EA 39.23 USD
77616-798 77686-143 77686-144
N-(9-Fluorenylmethoxycarbonyl)-5-aminopentan-1-ol ≥95%
N-(9-Fluorenylmethoxycarbonyl)-5-aminopentan-1-ol

Precaución: for Research Use Only

Formula: C₂₀H₂₃NO₃
MDL Number: MFCD00377789
CAS Number: 209115-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR