tert-Butyl 4-(methylsulfonyl)piperidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A412781-1G A412781-250MG A412781-5G
77594-270EA 94.3 USD
77594-270 77746-835 77746-836
tert-Butyl 4-(methylsulfonyl)piperidine-1-carboxylate ≥98%
tert-Butyl 4-(methylsulfonyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄S
MW: 263.35 g/mol
MDL Number: MFCD24447854
CAS Number: 189205-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR