7-((3S,4S)-3-Methoxy-4-(methylamino)pyrrolidin-1-yl)-4-oxo-1-(thiazol-2-yl)-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid hydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Voreloxin HCl
A405214-10MG
A405214-1MG
A405214-5MG
77616-756EA
110.21
USD
77616-756
77616-758
77616-760
7-((3S,4S)-3-Methoxy-4-(methylamino)pyrrolidin-1-yl)-4-oxo-1-(thiazol-2-yl)-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid hydrochloride ≥98%
7-((3S,4S)-3-Methoxy-4-(methylamino)pyrrolidin-1-yl)-4-oxo-1-(thiazol-2-yl)-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₁₈H₂₀ClN₅O₄S |
MDL Number:
MFCD13185156 CAS Number: 175519-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...