5-Fluoro-6-methoxynicotinaldehyde ≥98%

Supplier: Ambeed
Warning

A412194-100MG A412194-250MG
77890-846EA 47 USD
77890-846 77890-847
5-Fluoro-6-methoxynicotinaldehyde ≥98%
5-Fluoro-6-methoxynicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD04112587
CAS Number: 884494-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR