2-Fluoro-4-methoxybenzoic acid ≥98%

Supplier: Ambeed
Warning

A404551-5G A404551-1G A404551-100G A404551-25G
77594-226EA 17.43 USD
77594-226 77672-976 77882-075 77882-076
2-Fluoro-4-methoxybenzoic acid ≥98%
2-Fluoro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MDL Number: MFCD00509373
CAS Number: 394-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR