1H-Pyrrolo[2,3-b]pyridine-6-carbonitrile ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Cyano-7-azaindole
A404521-250MG
A404521-1G
77684-144EA
26
USD
77684-144
77684-143
1H-Pyrrolo[2,3-b]pyridine-6-carbonitrile ≥97%
1H-Pyrrolo[2,3-b]pyridine-6-carbonitrile
Precaución: For research use only.
Formula:
C₈H₅N₃ MW: 143.15 g/mol |
MDL Number:
MFCD06659673 CAS Number: 189882-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...