Methyl-4,5-dibromo-2-thiophenecarboxylate ≥95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4,5-dibromothiophene-2-carboxylate

A404397-1G A404397-5G
77879-881EA 51 USD
77879-881 77879-882
Methyl-4,5-dibromo-2-thiophenecarboxylate ≥95%
Methyl-4,5-dibromo-2-thiophenecarboxylate

Precaución: for Research Use Only

Formula: C₆H₄Br₂O₂S
MDL Number: MFCD00831688
CAS Number: 62224-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR