5-Fluoro-2-methoxy-3-pyridineboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (5-Fluoro-2-methoxypyridin-3-yl)boronic acid

A404289-1G A404289-250MG A404289-5G
77878-146EA 135 USD
77878-146 77878-147 77878-148
5-Fluoro-2-methoxy-3-pyridineboronic acid ≥98%
5-Fluoro-2-methoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₃
MDL Number: MFCD07368239
CAS Number: 957120-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR