Ethyl-1-boc-5-oxoazepane-4-carboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-tert-Butyl 4-ethyl 5-oxoazepane-1,4-dicarboxylate

A411747-25G A411747-5G
77668-862EA 89 USD
77668-862 77668-864
Ethyl-1-boc-5-oxoazepane-4-carboxylate ≥98%
Ethyl-1-boc-5-oxoazepane-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₅
MW: 285.336 g/mol
MDL Number: MFCD03788451
CAS Number: 141642-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR