3-Acetamido-3-(4-fluorophenyl)propanoic acid ≥97%

Supplier: Ambeed
Warning

A403949-5G A403949-250MG A403949-1G
77668-842EA 216 USD
77668-842 77879-879 77879-878
3-Acetamido-3-(4-fluorophenyl)propanoic acid ≥97%
3-Acetamido-3-(4-fluorophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO₃
MW: 225.21 g/mol
MDL Number: MFCD01366670
CAS Number: 332052-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR