3-Isobutylisoxazole-5-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A403828-250MG A403828-1G
77601-770EA 349.53 USD
77601-770 77882-074
3-Isobutylisoxazole-5-carboxylic acid ≥95%
3-Isobutylisoxazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₁NO₃
MDL Number: MFCD08059886
CAS Number: 910321-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR