(3-Fluoro-6-methylpyridin-2-yl)boronic acid 98+%

Supplier: Ambeed
Warning

A403607-1G A403607-5G
77892-832EA 209 USD
77892-832 77892-833
(3-Fluoro-6-methylpyridin-2-yl)boronic acid 98+%
(3-Fluoro-6-methylpyridin-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₂
MDL Number: MFCD28400996
CAS Number: 2093106-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR